Learn More
Aviva Systems Biology Corp Igf2bp1 Antibody - N-terminal region (ARP58385_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58385_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Igf2bp1. It was validated on Western Blot. . Alternate names: AL024068, AW549074, CRD-BP, Crdbp, D030026A21Rik, D11Moh40e, D11Moh45, IMP-1, Neilsen, Zbp1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-470