Learn More
Aviva Systems Biology Corp IGF2BP1 Antibody - N-terminal region (ARP40658_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40658_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGF2BP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IGF2BP1. Alternate names: CRD-BP, CRDBP, IMP-1, IMP1, VIon ChannelKZ1, ZBP1, VICKZ1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53220