Learn More
Aviva Systems Biology Corp IGF2BP2 Antibody - middle region (ARP40662_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40662_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGF2BP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IGF2BP2. Alternate names: IMP-2, IMP2, VICKZ2, p62ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-57577