Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IGF2BP2 Human T7-H 50ug
SDP

Catalog No. 5000196975
Encompass_Preferred

Recombinant human IGF2BP2 protein (residues M1-T220) supplied as a lyophilized powder. Expressed in E. coli with an N-terminal T7 tag and a C-terminal 6xHis tag, the protein is intended for biochemical assays, antibody validation, and binding studies. Purity is ≥96% by reducing SDS-PAGE and the approximate molecular weight is 31.0 kDa.

  • Lyophilized recombinant protein format for long-term storage.
  • Contains N-terminal T7 and C-terminal 6xHis affinity tags for detection and purification.
  • Sequence corresponds to human IGF2BP2 residues M1-T220 (accession Q9Y6M1-2).
  • Purity ≥96% as determined by reducing SDS-PAGE.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O; aliquot and freeze at -20°C or -80°C for extended storage.
  • Approximate molecular weight 31.0 kDa.
  • Suitable for biochemical assays, antibody validation, and binding studies.

Catalog No. 50-001-96975 Supplier Medchemexpress LLC Supplier No. HYP7095050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human IGF2BP2 protein (residues M1-T220) supplied as a lyophilized powder. Expressed in E. coli with an N-terminal T7 tag and a C-terminal 6xHis tag, the protein is intended for biochemical assays, antibody validation, and binding studies. Purity is ≥96% by reducing SDS-PAGE and the approximate molecular weight is 31.0 kDa.

  • Lyophilized recombinant protein format for long-term storage.
  • Contains N-terminal T7 and C-terminal 6xHis affinity tags for detection and purification.
  • Sequence corresponds to human IGF2BP2 residues M1-T220 (accession Q9Y6M1-2).
  • Purity ≥96% as determined by reducing SDS-PAGE.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O; aliquot and freeze at -20°C or -80°C for extended storage.
  • Approximate molecular weight 31.0 kDa.
  • Suitable for biochemical assays, antibody validation, and binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.