Learn More
Cayman Chemical IGF2BP2 Long Isoformhumn50ug
SDP
Catalog No. 502944151
Encompass_Preferred
Source: Recombinant human N-terminal His- and GST-tagged IGF2BP2 long isoform expressed in insect cells • Amino acids: 1-599 (full length) • MW: 94 kDa
Catalog No. 50-294-4151 Supplier Cayman Chemical Supplier No. 4153350 UG
Source: Recombinant human N-terminal His- and GST-tagged IGF2BP2 long isoform expressed in insect cells • Amino acids: 1-599 (full length) • MW: 94 kDa