Learn More
Proteintech Group Inc IGF2BP2 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 116011AP150UL
Encompass_Preferred
The IGF2BP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IGF2BP2. This antibody recognizes human, mouse, rat antigen. The IGF2BP2 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Catalog No. 50-225-6373