Learn More
Aviva Systems Biology Corp Igf2bp3 Antibody - middle region (ARP40660_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40660_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Igf2bp3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2610101N11Rik, AA522010, AL022933, AU045931, IMP-3, IMP3, Koc13, MGC61294, Neilsen, mimp3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-284-886