Learn More
Aviva Systems Biology Corp IGFALS Antibody - middle region (ARP42206_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42206_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGFALS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IGFALS. Alternate names: ALSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-58141