Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IGFBP-2 ELISA Kit

Catalog No. EMIGFBP2X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMIGFBP2X5 5 x 96 Tests
EMIGFBP2 96 Tests
EMIGFBP2X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EMIGFBP2X5 Supplier Invitrogen™ Supplier No. EMIGFBP2X5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Mouse IGF Binding Protein 2 (Ms IGFBP-2) ELISA quantitates Ms IGFBP-2 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IGFBP-2. Principle of the method The Mouse IGFBP-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IGFBP2 (insulin-like growth factor binding protein 2) is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P47877
Assay Range 0.6-600 ng/mL
Assay Sensitivity 600 pg/mL
Calibration Range 0.6 ng/mL to 600 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
Sufficient For 5 x 96 assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI255832; BP2; BRL-BP; IBP2; IBP-2; IGF binding protein 2; IGF-binding protein 2; IGFBP; IGFBP2; IGFBP-2; IGF-BP53; ILGFBPA; Insulin like growth factor binding protein; insulin like growth factor binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36kDa; insulin-like growth factor-binding protein 2; mIGFBP-2; OTTHUMP00000207753AI255832; BP2; BRL-BP; IBP2; IBP-2; IGF binding protein 2; IGF-binding protein 2; IGFBP; IGFBP2; IGFBP-2; IGF-BP53; ILGFBPA; Insulin like growth factor binding protein; insulin like growth factor binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36kDa; insulin-like growth factor-binding protein 2; mIGFBP-2; OTTHUMP00000207753; AI255832; IBP-2; Igfbp-2; mIGFBP-2
Gene ID (Entrez) 16008
Gene Symbol IGFBP2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases IBP-2, IGF-binding protein 2, IGFBP-2, mIGFBP-2
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Development
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym AI255832, IBP-2, Igfbp-2, mIGFBP-2
Target IGFBP2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.