Learn More
ACROBiosystems ACROBiosystems Human IGFBP-4 Protein, His Tag
Supplier:  ACROBiosystems IG4H52E450UG
Encompass_Preferred
Recombinant Protein;50UG;Human IGFBP-4, His Tag (IG4-H52E4) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Glu 258 (Accession # AAH16041).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.9 kDa. The protein migrates as 33-37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IGFBP-4, His Tag (Cat. No. IG4-H52E4) at 2 μg/mL (100 μL/well) can bind Human IGF-I, Fc Tag (Cat. No. IG1-H4269) with a linear range of 4-31 ng/mL (QC tested).
Catalog No. 50-195-7299