Learn More
Ansh Labs Ansh Labs Intact IGFBP-4 ELISA kit, 96-well colorimetric assay. Quantitative detection in serum and other biological fluids in human, equine, canine, bovine, caprine, mouse and rat. Dynamic range of 1.5-96.16 ng/mL & sensitivity of 0.669 ng/mL.
SDP
Supplier:  Ansh Labs AL128
Encompass
The Intact IGFBP-4 ELISA kit provides materials for the quantitative measurement of IGFBP-4 (Insulin-like Growth Factor Binding Protein 4) in human serum and other biological fluids, with species reactivity to equine, canine, bovine, caprine, mouse and rat samples. The kit offers an analytical range of 1.5-96.16 ng/mL and a sensitivity of 0.669 ng/mL, using 25 uL of serum. The kit is an HRP-based ELISA with colorimetric detection by dual wavelength absorbance at 450 nm and 630 nm as reference filter and has an assay time of 1.5 hours. For Research Use Only.
Catalog No. NC1406553