Learn More
Crystal Chem Inc IGFBP-6 ELISA, 96-wells
SDP
Supplier:  Crystal Chem Inc 80595
Encompass
The IGFBP-6 ELISA is used to quantify levels of human IGFBP-6 in serum, plasma, fluids, and cell culture using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL. Dynamic range of 0.1 - 10 ng/mL. High sensitivity of 0.03 ng/mL.
Catalog No. 50-194-7758