Learn More
BioVendor, LLC Igfbp-6 Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC REH007R
Encompass
The BioVendor Human IGFBP-6 ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IGFBP-6 in serum, plasma, and cell culture supernatants. This assay employs an antibody specific for human IGFBP-6 coated on a 96well plate. Standards and samples are pipetted into the wells and IGFBP-6 present in a sample is bound to the wells by the immobilized antibody. The wells are washed and biotinylated anti-human IGFBP-6 antibody is added. After washing away unbound biotinylated antibody, HRP-conjugated streptavidin is pipetted to the wells. The wells are again washed, a TMB substrate solution is added to the wells and color develops in proportion to the amount of IGFBP-6 bound. The Stop Solution changes the color from blue to yellow, and the intensity of the color is measured at 450 nm. One kit (96 wells). Calibration range: 82.3-60,000pg/ml. Limit of detection: 150pg/ml. Other names: insulin-like growth factor binding protein-6.
Catalog No. 50-240-6382