Learn More
BioVendor, LLC Igfbp-7 Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL015R
Encompass
IGFBP7 is a member of the insulin superfamily of growth promoting peptides, and is one of the most abundant and common growth factor polypeptides. IGFBPs are a class of secreted proteins that is able to interact with many ligands other than IGFs. Currently, IGFBP-7 has been used as an early diagnosis and prognostic marker for acute renal insufficiency (AKI). IGFBP-7 has also been proposed as a potential prognostic biomarker in heart failure (HF). It provided prognostic information incremental to clinical variables, NT-proBNP, and hsTnT. One kit (96 wells). Type: Sandwich ELISA. Sample type: serum, plasma, urine. Sample requirement: 2 ¿l/well. Calibration range: 0.125-4ng/ml. Limit of Detection: 31pg/ml. Other names: Insulin-like growth factor binding protein-7, IBP-7, Mac25, IGF binding protein related protein-1 (IGFBPrP1).
Catalog No. 50-240-6359