Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Igfbp3 (Mouse) ELISA Kit

Catalog No. 89101003 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-003 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-028-523 CCS, Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-003 Supplier Abnova™ Supplier No. KA3324
Add to Cart
Add to Cart

In-vitro ELISA for the quantitative measurement of Mouse Igfbp3

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Igfbp3 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Igfbp3.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-mouse IGFBP-3 antibody, Lyophilized IGFBP-3 standards (10ng x 2), Sample diluent buffer (30mL), Biotin conjugated anti-mouse IGFBP-3 antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (130uL), Antibody diluent buffer (12mL), ABC diluent buffer (12mL), TMB color developing agent (10mL), Stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type IGFBP3 (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.