Learn More
Aviva Systems Biology Corp IGFBP4 Antibody - middle region (ARP45685_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45685_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGFBP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IGFBP4. Alternate names: BP-4, HT29-IGFBP, IBP4, IGFBP-4ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-53451