Learn More
Aviva Systems Biology Corp IGFBP7 Antibody - C-terminal region (ARP48174_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48174_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGFBP7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human IGFBP7. Alternate names: FSTL2, IGFBP-7, IGFBP-7v, MAC25, PSF, AGM, TAF, IBP-7, RAMSVPS, IGFBPRP1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-53542