Learn More
ACROBiosystems Recombinant Protein;Human IGFBP-7 Protein;HEK293;1MG;IG7,IBP7,IGFBP7,IBP-7,IGFBP-7,IGFBP-rP1,TAF,MAC25,PSF
Supplier:  ACROBiosystems IG7H52401MG
Encompass_Preferred
Recombinant Protein;Human IGFBP-7 Protein;1MG;Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession # NP_001544).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8936