Learn More
ACROBiosystems Recombinant Protein;Mouse IGF-I R / CD221 Protein;HEK293;100UG;IGR,IGF1R,CD221,IGFIR,IGFR,JTK13,IGF-IR,IGF-1R,JTK-13,MGC142170,MGC142172,MGC18216,IGF-I R
Catalog No. 501657665
Encompass_Preferred
Recombinant Protein;Mouse IGF-I R / CD221 Protein;100UG;Mouse IGF-I R (IGR-M5223) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - His 936 (Accession # Q60751-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 105.6 kDa, 80.7 kDa, 24.9 kDa. The protein migrates as 45-50 kDa, 96-115 kDa and 120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse IGF-I R (Cat. No. IGR-M5223) at 5 μg/mL (100 μL/well) can bind Human IGF-I, Fc Tag (Cat. No. IG1-H4269) with a linear range of 0.04-0.6 μg/mL (QC tested).
Catalog No. 50-165-7665 Supplier ACROBiosystems Supplier No. IGRM5223100UG
Recombinant Protein;Mouse IGF-I R / CD221 Protein;100UG;Mouse IGF-I R (IGR-M5223) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - His 936 (Accession # Q60751-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 105.6 kDa, 80.7 kDa, 24.9 kDa. The protein migrates as 45-50 kDa, 96-115 kDa and 120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse IGF-I R (Cat. No. IGR-M5223) at 5 μg/mL (100 μL/well) can bind Human IGF-I, Fc Tag (Cat. No. IG1-H4269) with a linear range of 0.04-0.6 μg/mL (QC tested).