Learn More
ACROBiosystems Recombinant Protein;Rat IGF-I R / CD221 Protein;HEK293;1MG;IGR,IGF1R,CD221,IGFIR,IGFR,JTK13,IGF-IR,IGF-1R,JTK-13,MGC142170,MGC142172,MGC18216,IGF-I R
Catalog No. 501657662
Encompass_Preferred
Recombinant Protein;Rat IGF-I R / CD221 Protein;1MG;Rat IGF-I R (IGR-R5224) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - His 936 (Accession # P24062-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 105.5 kDa, 81.3 kDa, 24.2 kDa. The protein migrates as 45-50 kDa, 96-115 kDa and 120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rat IGF-I R (Cat. No. IGR-R5224) at 5 μg/mL (100 μL/well) can bind Human IGF-I, Fc Tag (Cat. No. IG1-H4269) with a linear range of 0.01-0.313 μg/mL (QC tested).
Catalog No. 50-165-7662 Supplier ACROBiosystems Supplier No. IGRR52241MG
Recombinant Protein;Rat IGF-I R / CD221 Protein;1MG;Rat IGF-I R (IGR-R5224) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - His 936 (Accession # P24062-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 105.5 kDa, 81.3 kDa, 24.2 kDa. The protein migrates as 45-50 kDa, 96-115 kDa and 120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rat IGF-I R (Cat. No. IGR-R5224) at 5 μg/mL (100 μL/well) can bind Human IGF-I, Fc Tag (Cat. No. IG1-H4269) with a linear range of 0.01-0.313 μg/mL (QC tested).