Learn More
AdipoGen IGFL (mouse) (rec.),Protein. Mouse IGFL
SDP
Supplier:  AdipoGen AG40B0091C010
Encompass_Preferred
IGFL (mouse) (rec.), AdipoGen Life Sciences. Protein. Mouse IGFL (aa 25-140) is fused at the N-terminus to a FLAG?-tag.  Lyophilized. Contains PBS.
Catalog No. 50-114-6126