Learn More
AdipoGen IGFL1 human rec
SDP
Supplier:  AdipoGen AG40B0244C010
Encompass_Preferred
Recombinant protein. Human IGFL1 aa 25-110 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Lyophilized. Contains PBS. The Insulin-growth factor-like gene family is a new family of proteins consisting of four proteins in humans IGFL1 to 4 and one in mice mIGFL. mIGFL is expressed in normal skin in mice and further upregulated during inflammation responses in the skin or after skin wounding. In humans only IGFL1 expression is increased in psoriatic skin samples. mIGFL and human IGFL1 and 3 interact with specificity and high affinity to a novel receptor named IGF-like family receptor 1 formerly TMEM-149. Analysis of the amino acid sequence of IGFLR1 indicated that this receptor is likely a novel member of the TNF-R family. IGFLR1 is expressed most abundantly on mouse T cells, suggesting that mIGFL and IGFL1 produced in the skin may potentially exert regulatory functions on T cell responses.
Catalog No. 50-256-2813