Learn More
AdipoGen IGFL3 (human) (rec.),Protein. Human IGFL3
SDP
Catalog No. 501175375
Encompass_Preferred
IGFL3 (human) (rec.), AdipoGen Life Sciences. Protein. Human IGFL3 (aa 25-125) is fused at the N-terminus to a FLAG¿-tag.  Lyophilized. Contains PBS.
Catalog No. 50-117-5375 Supplier AdipoGen Supplier No. AG40B0090C010
IGFL3 (human) (rec.), AdipoGen Life Sciences. Protein. Human IGFL3 (aa 25-125) is fused at the N-terminus to a FLAG¿-tag.  Lyophilized. Contains PBS.