Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3500MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: IG217 + IG266. Clonality: Monoclonal. Host: Mouse. Isotype: IIgG2a kappa. Positive Control: 293T, Raji or hPBL cells. Tonsil or Spleen. Cellular Staining: Cytoplasm, Cell Surface, Secreted. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Purified human IgG. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~75kDa.Chromosome: 14q32.33. Gene Symbol: IGHG. Gene ID: 3503.
Catalog No. 50-188-4798