Learn More
Innovative Research Inc Human IgG Affinity Purified 1000mg
Supplier:  Innovative Research Inc IHUIGGAP1000MG
Encompass_Preferred
Human IgG Affinity Purified from Innovative Research has been purified from normal Human Serum using Protein A affinity chromatography. This is a frozen liquid buffered in 0.02M Sodium Phosphate; 0.15M NaCl; pH 7.2, with a concentration of 10.0 mg/ml and a purity of >95%.
Catalog No. 50-203-6434