Learn More
Roche Diagnostics IgG Cedex Bio Instrument
Supplier:  Roche Diagnostics 06681743001
Encompass
This product is intended for quantitative determination of human Immunoglobin G (IgG) in aqueous solution using the Cedex Bio Instrument.
Catalog No. 50-589-434