Learn More
Pel Freez Biologicals IgG Depleted Baby Rabbit Complement, 100ml
Supplier:  Pel Freez Biologicals 31065100
Encompass
Complement is produced under controlled conditions from large pools of New Zealand White rabbits that are 21-28 days old then depleted of IgG antibodies further reducing heterophile antibody activity. Useful in serum bactericidal and opsonophagocytic assays.
Catalog No. NC3593139