Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter IgG Determinant

Catalog No. 21280052 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
168 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-280-052 168 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-052 Supplier Beckman Coulter Supplier No. OSR61172
Add to Cart
Add to Cart
This item is not returnable. View return policy

IgG assay offers exceptional clinical quality for Proteins/Serology testing.

  • Our broad and growing portfolio of high-performing Six Sigma assays is compatible with AU Chemistry Analyzers - and it offers exceptional clinical quality
  • Load it and leave it with liquid ready-to-use reagents
  • Trust results with standardized test menus, assay protocols and reference ranges providing commutable patient results across AU analyzers
TRUSTED_SUSTAINABILITY

Specifications

Type Reagent
For Use With (Equipment) Beckman Coulter Analyzer
Quantity 168 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.