Learn More
Crystal Chem Inc IgG ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80720
Encompass
The IgG ELISA is used to quantify levels of human IgG in serum and plasma using only a 2 µL sample volume. Highlights: Kit uses sample size of only 2 µL. High sensitivity of 1.8 ng/mL. Dynamic range of 15.6 - 500 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7840