Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, F(ab')2 Fragment Goat Anti-Rabbit, Affinity Purified, Secondary Antibody, MP Biomedicals™

Catalog No. MP08670391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08670391 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08670391 Supplier MP Biomedicals, Inc Supplier No. 08670391
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Anti-Rabbit Secondary AntibodyAffinity Purified

  • Goat anti-rabbit IgG, F(ab')2 fragment is used in ELISA, Immunohistochemistry technique, and in western blot.
  • Immunoglobulin G (IgG) are antibody molecules.
  • Each IgG is composed of four peptide chains - two heavy chains and two light chains.
  • Each IgG has two antigen binding sites.
  • Other Immunoglobulins may be described in terms of polymers with the IgG structure considered the monomer.
  • The Fab region, (fragment, antigen binding), is the binding region of the antibody.
TRUSTED_SUSTAINABILITY

Specifications

Applications ELISA, Western Blot
Concentration Antibody Concentration: 2.3mg/mL
Conjugate Unconjugated
Description Goat anti-rabbit IgG (Fab')2
Formulation supplied in 0.01M sodium phosphate, 0.25M NaCl, pH 7.6. This product does not contain any preservatives
Host Species Goat
Immunogen Goat anti-rabbit IgG (Fab')2 was raised ingoat using rabbit IgG F(ab')2 fragment as the immunogen
Purification Method Affinity Purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at 2-8°C. After opening dilute only enough antibody for a single day's use. Store the remainder at 2-8°C under sterile conditions.
Form Liquid
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.