Learn More
Athens Research And Technology Inc IGG FAB FRAGMENT HU PLASMA10MG
Supplier:  Athens Research And Technology Inc 1616090707FAB10MG
Encompass_Preferred
Immunoglobulin G, Fab Fragment, Human Plasma: 2x5 MG; Frozen in 20 mM phosphate, pH 7.4, with 150 mM NaCl and 0.05% NaN3; MW 55,000; EC=1.35; Purity: > =95% by SDS-PAGE. No reaction by IEP to antisera to human IgG-fc. Native;
Catalog No. 50-489-787