Learn More
Cayman Chemical IgG Fabmouse Monclonal n 50ug
SDP
Catalog No. 502950084
Encompass_Preferred
Immunogen: Mouse IgG • Host: Goat • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG1, IgG2a, IgG2b, IgG3; (-) Mouse IgM, IgA, IgE (-) Human, rabbit, rat IgG • Application: ELISA
Catalog No. 50-295-0084 Supplier Cayman Chemical Supplier No. 3236150 UG
Immunogen: Mouse IgG • Host: Goat • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG1, IgG2a, IgG2b, IgG3; (-) Mouse IgM, IgA, IgE (-) Human, rabbit, rat IgG • Application: ELISA