Learn More
Cayman Chemical IgG FabrabbIt MoncLNal A 100ug
SDP
Supplier:  Cayman Chemical 32099100 UG
Encompass_Preferred
Immunogen: Rabbit IgG • Host: Goat • Species Reactivity: (+) Rabbit • Cross Reactivity: (-) Human, mouse, and rat IgG • Applications: ELISA, WB • MW: ~150 kDa for non-reduced IgG
Catalog No. 50-294-7791