Learn More
Specifications
Specifications
| Antigen | IgG Fc |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Goat anti-rabbit IgG recognizes rabbit IgG F(c) fragment. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | In 10mM sodium phosphate, 150mM NaCl, pH 7.2 |
| Host Species | Goat |
| Immunogen | Rabbit IgG F(c) fragment |
| Purification Method | This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. |
| Quantity | 50 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.