Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Fc Goat anti-Rat, Polyclonal , Abnova™

Catalog No. 89946209 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-209 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-946-209 Supplier Abnova Corporation Supplier No. PAB29754
Add to Cart
Add to Cart

Goat polyclonal antibody raised against rat IgG Fc.

Specifications

Antigen IgG Fc
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against rat IgG Fc.
Dilution ELISA (2 ug/mL) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Goat
Immunogen Rat IgG Fc fragment.
Purification Method Antigen Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity It reacts only with Rat IgG by immunodiffusion and IEP techniques. This does not imply an equal balance of reactivity to all IgG subclasses.
Target Species Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.