Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ IgG(Fc specific), Goat anti-Rabbit, AP, Polyclonal,

Catalog No. 692653MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
40 μL
Conjugate:
Alkaline Phosphatase (AP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
69-265-3MI 40 μL Alkaline Phosphatase (AP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-265-3MI Supplier MilliporeSigma™ Supplier No. 692653
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG(Fc specific)
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Host Species Goat
Quantity 40 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage -20°C
Isotype IgG

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.