Learn More
Diagnostic Biosystems IGG-FITC IgG-FITC
Supplier:  Diagnostic Biosystems F008
Encompass_Preferred
This antibody reacts with the gamma chain of human IgG. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.
Catalog No. 50-240-7528