Learn More
Sigma Aldrich Fine Chemicals Biosciences IgG-fITC from human serum
Catalog No. 502900024
Encompass
IgGs are glycoprotein antibodies that regulate immunological defense mechanisms such as phagocytosis and inflammatory responses . IgG also modulates complement-mediated cell lysis . IgG-fITC from human serum does not react with purified human IgA IgG IgM and Bence Jonesº and lambda myeloma proteins. Additionally the product does not react with serum from rat goat rabbit guinea pig and mouse.
Catalog No. 50-290-0024 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F96361ML
IgGs are glycoprotein antibodies that regulate immunological defense mechanisms such as phagocytosis and inflammatory responses . IgG also modulates complement-mediated cell lysis . IgG-fITC from human serum does not react with purified human IgA IgG IgM and Bence Jonesº and lambda myeloma proteins. Additionally the product does not react with serum from rat goat rabbit guinea pig and mouse.