Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Fraction Goat anti-Mouse, FITC, Polyclonal, MP Biomedicals

Catalog No. MP855503 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP855503 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP855503 Supplier MP Biomedicals, Inc Supplier No. 0855503
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG Fraction Polyclonal antibody specifically detects IgG Fraction in Mouse samples. It is validated for Immunohistochemistry (Frozen) and Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Fraction
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Total Protein: 14.9mg/mL; Antibody Protein: 3.0mg/mL
Conjugate FITC
Host Species Goat
Immunogen Mouse IgG
Purification Method Conjugated protein is purified by gel filtration. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22μm filter, vialed, and lyophilized.
Quantity 2 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store lyophilized product at 2° to 8°C. Store reconstituted material no longer than 4 weeks at 2° to 8°C. For long-term storage, aliquot and freeze reconstituted product at -20°C or lower.
Form Lyophilized
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.