Learn More
Jackson Immuno Research Labs Peroxidase IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, light chain specific. .5ml
SDP
Catalog No. NC9364937
Encompass_Preferred
Peroxidase-IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific (min X Bov,Gt,Arm Hms,Hrs,Hu,Ms,Rat,Shp Ig)
Catalog No. NC9364937 Supplier Jackson Immuno Research Labs Supplier No. 211032171
Peroxidase-IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific (min X Bov,Gt,Arm Hms,Hrs,Hu,Ms,Rat,Shp Ig)