Learn More
Innovative Research Inc Mouse IgG Fractionated Purified Lyophilized 1gm
Supplier:  Innovative Research Inc IMSIGGGFLY1GM
Encompass_Preferred
Mouse IgG Fractionated Purified Lyophilized from Innovative Research has been purified from mouse serum by fractionation. This is a lyophilized powder buffered in 10mM sodium phosphate, 150mM sodium chloride pH 7.2, with a purity of greater than 97% as determined by SDS-PAGE analysis.
Catalog No. 50-203-6511