Learn More
Sigma Aldrich Fine Chemicals Biosciences IgG from bovine serum reag10MG
Catalog No. 502900894
Encompass
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Bovine IgG is purified from pooled normal bovine serum by fractionation and ion-exchange chromatography.
Catalog No. 50-290-0894 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I550610MG
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Bovine IgG is purified from pooled normal bovine serum by fractionation and ion-exchange chromatography.