Learn More
Innovative Research Inc Human IgG from Covid-19 Convalescent Plasma Fractionated Purified Lyophilized 50ug
Supplier:  Innovative Research Inc IHUCOVIGGGFLY50UG
Encompass_Preferred
Human IgG from Covid-19 Convalescent Plasma Fractionated Purified Lyophilized from Innovative Research has been purified by immobilized Protein A from pooled sera collected from convalescent COVID-19 patients. This is a Lyophilized powder buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6.
Catalog No. 50-203-6526