Learn More
Sigma Aldrich Fine Chemicals Biosciences IgG from goat serum reagen50MG
Catalog No. 502904255
Encompass
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Goat IgG is purified from normal goat serum by fractionation and ion-exchange chromatography.
Catalog No. 50-290-4255 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I525650MG
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Goat IgG is purified from normal goat serum by fractionation and ion-exchange chromatography.