Learn More
Sigma Aldrich Fine Chemicals Biosciences IgG from mouse serum techn10MG
Catalog No. 502900342
Encompass
Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. A immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Mouse consists of five immunoglobulin classes- IgM IgG IgA IgD and IgE. Mouse IgG is further divided into five classes- IgG1 IgG2a IgG2b and IgG3.
Catalog No. 50-290-0342 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I876510MG
Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. A immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Mouse consists of five immunoglobulin classes- IgM IgG IgA IgD and IgE. Mouse IgG is further divided into five classes- IgG1 IgG2a IgG2b and IgG3.