Learn More
Sigma Aldrich Fine Chemicals Biosciences IgG from rabbit serum reag10MG
Catalog No. 502900895
Encompass
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Rabbit IgG is purified from normal rabbit serum by fractionation and ion-exchange chromatography.
Catalog No. 50-290-0895 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I500610MG
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Rabbit IgG is purified from normal rabbit serum by fractionation and ion-exchange chromatography.