Learn More
Sigma Aldrich Fine Chemicals Biosciences IgG from rabbit serum reag50MG
Catalog No. 502904378
Encompass
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Rabbit IgG is purified from normal rabbit serum by fractionation and ion-exchange chromatography.
Catalog No. 50-290-4378 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I500650MG
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Rabbit IgG is purified from normal rabbit serum by fractionation and ion-exchange chromatography.