Learn More
AdipoGen IgG guinea pig pAb HRP
SDP
Catalog No. 502563098
Encompass_Preferred
Polyclonal Antibody. Reacts with the heavy and light chains of guinea pig IgG. Shows minimal cross-reactivity with bovine, chicken, goat, Syrian hamster, horse, human, mouse, rabbit, rat and sheep immunoglobulins. Source/Host Donkey. Applications ELISA, ICC, IHC, WB. Liquid. In PBS and BSA. The anti-IgG guinea pig, pAb HRP reacts with the whole molecule guinea pig IgG and also with the light chains of other guinea pig immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. The antibody shows minimal cross-reaction with human, mouse, rat, goat, bovine, chicken, Syrian hamster, horse, rabbit and sheep serum proteins, but it may cross-react with immunoglobulins from other species.
Catalog No. 50-256-3098 Supplier AdipoGen Supplier No. AG29B0008ER050
Polyclonal Antibody. Reacts with the heavy and light chains of guinea pig IgG. Shows minimal cross-reactivity with bovine, chicken, goat, Syrian hamster, horse, human, mouse, rabbit, rat and sheep immunoglobulins. Source/Host Donkey. Applications ELISA, ICC, IHC, WB. Liquid. In PBS and BSA. The anti-IgG guinea pig, pAb HRP reacts with the whole molecule guinea pig IgG and also with the light chains of other guinea pig immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. The antibody shows minimal cross-reaction with human, mouse, rat, goat, bovine, chicken, Syrian hamster, horse, rabbit and sheep serum proteins, but it may cross-react with immunoglobulins from other species.