Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (H, L) Chicken anti-Cotton Rat, Polyclonal , Abnova™

Catalog No. 89939470 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-939-470 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-939-470 Supplier Abnova Corporation Supplier No. PAB29752
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against cotton rat IgG (H&L).

Specifications

Antigen IgG (H, L)
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against cotton rat IgG (H&L).
Dilution ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Chicken
Immunogen Cotton rat IgG (H&L) whole molecule.
Purification Method Antigen Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity The affinity purified anti Cotton rat IgG reacts with immunoglobulins and the papain derived Fc and Fab fragments by IEP and gel diffusion techniques. It also reacts with rat, gerbil and hamster IgG.
Target Species Cotton Rat
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.