Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (H&L) Goat anti-Rabbit, DyLight™488, Abnova™

Catalog No. 89336025 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-336-025 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-336-025 Supplier Abnova Corporation Supplier No. MAB19500
Add to Cart
Add to Cart

Goat Monoclonal Antibody.

IgG (H&L) Monoclonal antibody specifically detects in Rabbit samples. It is validated for ICC, IHC-P, Flow Cyt.

Specifications

Antigen IgG (H&L)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Conjugate DyLight 488
Host Species Goat
Immunogen Rabbit IgG whole molecule
Quantity 1 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.